missing translation for 'onlineSavingsMsg'
Learn More

CD131, Biotin, clone: 1C1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-1319-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15537326

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The 1C1 monoclonal antibody reacts with the human CD131 molecule, also known as the common β subunit (βC). The common β subunit associates with the specific α subunits of IL-3 receptor, IL-5 receptor and GM-CSF receptor to form high affinity receptors for these cytokines. These cytokine receptors are expressed by neutrophils, eosinophils, monocytes, endothelial cells, fibroblasts and hematopoietic progenitor cells and play a crucial role in growth/activation of eosinophils and in the inflammatory response.
Specifications

Specifications

CD131
1C1
Biotin
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
common beta subunit, Colony Stimulating Factor 2 Receptor beta
IgG1, kappa
RUO
Primary
Human
Videos

For Research Use Only.