missing translation for 'onlineSavingsMsg'
Learn More

CD130, PE, clone: KGP130, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1302-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15536876

  • £62.31 / 25µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The KGP130 monoclonal antibody reacts with mouse gp130, also known as CD130. gp130 is a 130 kD Type-I transmembrane glycoprotein containing a 597 amino acid extracellular domain, a single transmembrane domain, and a 277 amino acid cytoplasmic domain. gp130 is a subunit of several heterodimeric cell-surface receptors, including receptors for IL-6, IL-11, IL-27, Oncostatin M, and Leukemia Inhibitory Factor (LIF). The gp130 protein has also been found to exist in a soluble form, which is capable of inhibiting IL-6 signaling. gp130 is expressed mainly on T cells, monocytes, endothelial cells, activated B cells, and plasma cells, and is expressed at lower levels on most leukocytes and epithelial cells. In response to ligand binding, gp130 becomes tyrosine phosphorylated, leading to activation of several signaling pathways including the PI3 kinase, Ras-MAPk and Stat pathways. Preincubation of mouse splenic T cells with IL-6 is capable of blocking the interaction of KGP130 with gp130, suggesting that the KGP130 binding site lies near the IL-6 interaction domain. Reactivity of KGP130 towards human gp130 has not been observed.Applications Reported: This KGP130 antibody has been reported for use in flow cytometric analysis.Applications Tested: This KGP130 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD130 (gp130)
KGP130
PE
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
gp130, IL-6 Receptor beta, IL-6RB
IgG2a
RUO
Primary
Murine
Videos