missing translation for 'onlineSavingsMsg'
Learn More

CD130, Biotin, clone: AN-G30, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-1308-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15229719

  • £62.63 / 25µg


Estimated Shipment date
from Supplier 01-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The AN-G30 monoclonal antibody reacts with human gp130, also known as CD130. gp130 is the common receptor subunit for the IL-6 family of cytokines, including IL-6, IL-11, IL-27, Oncostatin M, and LIF. gp130 is expressed on numerous cell types, including T cells, mast cells, and epithelial cells. Soluble forms of gp130 have been isolated from human serum samples. Upon ligand binding, gp130 signals via the JAK-STAT, MAPK, and PI3 kinase pathwaysApplications Reported: This AN-G30 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB).Applications Tested: This AN-G30 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD130 (gp130)
AN-G30
Biotin
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CD130, gp130, IL-6 Receptor beta, IL-6RB
IgG2a
RUO
Primary
Human
Videos