missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD13 Monoclonal Antibody (TUK 1), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD1301
Code : 50
Additional Details : Weight : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD13 Monoclonal antibody specifically detects CD13 in Human samples. It is validated for Flow Cytometry.Specifications
CD13 | |
Monoclonal | |
FITC | |
ANPEP | |
alanyl (membrane) aminopeptidase; alanyl (membrane) aminopeptidase (aminopeptidase N, aminopeptidase M, microsomal aminopeptidase, CD13, p150); alanyl aminopeptidase; alanyl aminopeptidase, membrane; aminopeptidase; aminopeptidase Ey; Aminopeptidase M; aminopeptidase N; aminopeptidase N.; aminopeptidase N/CD13; Anpep; APDE; Apm; AP-M; APN; AP-N; bAPN; cAPN; Cd13; cluster of differentiation antigen 13 (CD13); fAPN; gp130; gp150; h AP-N; hAPN; kidney aminopeptidase M; kidney Zn peptidase; KZP; Lap1; Lap-1; leucine arylaminopeptidase 1; LOW QUALITY PROTEIN: aminopeptidase N; membrane alanine aminiopeptidase; membrane alanine aminopeptidase; membrane alanyl aminopeptidase; membrane protein p161; Microsomal aminopeptidase; myeloid plasma membrane glycoprotein CD13; p150; pAPN; PEPN; porcine aminopeptidase N; rAPN; rbAPN; Unknown (protein for MGC:126949) | |
Mouse | |
purified | |
ASR | |
290 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
TUK 1 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P15144 | |
ANPEP | |
Human CD13 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |