missing translation for 'onlineSavingsMsg'
Learn More

CD13, clone: WM-15 (WM15), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0138-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15236817

  • £30.74 / 25µg


Estimated Shipment
28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The WM-15 monoclonal antibody reacts with human CD13, also known as aminopeptidase N. CD13 is a transmembrane ectoenzyme occurring on monocytes and granulocytes but not lymphocytes in blood. It is also reportedly on a wide variety of cells and in a wide variety of tissues, including endothelial cells, epithelial cells, fibroblasts, kidney proximal tubules, intestine, and placenta. CD13 is reported to have many different functions including degradation of enkephalins and endorphins, terminal degradation of peptides, amino acid scavenging, antigen processing, and adhesion and migration of cells.Applications Reported: The WM-15 (WM15) antibody) has been reported for use in flow cytometric analysis, and microscopy.Applications Tested: The WM-15 (WM15) antibody has been tested by flow cytometric analysis of lysed whole blood. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD13
WM-15 (WM15)
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Immunohistochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Alanyl Aminopeptidase (ANPEP), Aminopeptidase N (PEPN)
IgG1
RUO
Primary
Human
Videos