missing translation for 'onlineSavingsMsg'
Learn More

CD13 Mouse anti-Human, APC, Clone: WM-15 (WM15), eBioscience™

Mouse Monoclonal Antibody

£78.75 - £165.00

Specifications

Antigen CD13
Clone WM-15 (WM15)
Target Species Human
Conjugate APC
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15587876
View Documents
Affymetrix eBioscience
17-0138-41
25 Tests £78.75
25 tests View alternative products
Save Up to 
»
Estimated Shipment
27-10-2020
Please to purchase this item. Need a web account? Register with us today!
15597876
View Documents
Affymetrix eBioscience
17-0138-42
100 Tests £165.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment
27-10-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD13 (ANPEP) is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on various cell types. CD13 is a metalloprotease that preferentially catalyzes removal of neutral amino acids from small peptides, thus activating or inactivating bioactive peptides. CD13 also has a role in extracellular matrix degradation, antigen processing and signal transduction, is important in inflammatory responses, regulates intercellular contact, cell motility and vascularization. CD13 is expressed by granulocytes, monocytes, myeloid leukemia cells, and is involved in protection of leukemic cells against apoptosis and its expression associated with poor prognosis of carcinomas. CD13 functions as an aminopeptidase enzyme and is also alpha receptor for coronavirus.
Specifications

Specifications

CD13
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
WM-15 (WM15)
APC
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.