missing translation for 'onlineSavingsMsg'
Learn More

CD13 Mouse anti-Human, APC, Clone: WM-15 (WM15), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0138-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15587876

  • £78.75 / 25 tests

Estimated Shipment
05-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD13 (ANPEP) is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on various cell types. CD13 is a metalloprotease that preferentially catalyzes removal of neutral amino acids from small peptides, thus activating or inactivating bioactive peptides. CD13 also has a role in extracellular matrix degradation, antigen processing and signal transduction, is important in inflammatory responses, regulates intercellular contact, cell motility and vascularization. CD13 is expressed by granulocytes, monocytes, myeloid leukemia cells, and is involved in protection of leukemic cells against apoptosis and its expression associated with poor prognosis of carcinomas. CD13 functions as an aminopeptidase enzyme and is also alpha receptor for coronavirus.
Specifications

Specifications

CD13
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P15144
AP-M, AP-N, Apn, Cd13, P150, alanyl aminopeptidase, aminopeptidase M, aminopeptidase N, aminopeptidase N/CD13, membrane protein p161, microsomal aminopeptidase
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
WM-15 (WM15)
APC
ANPEP
Liquid
ANPEP
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
290
Videos
SDS
Documents

For Research Use Only.