missing translation for 'onlineSavingsMsg'
Learn More

CD127, PE, clone: eBioSB/199 (SB/199), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1273-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15278659

  • £68.27 / 50µg


EU Stock 2
Add to basket

Description

Description

Description: The SB/199 monoclonal antibody reacts with mouse CD127, the high affinity a subunit of the mouse IL-7 receptor. IL-7 receptor a chain is expressed by immature B cells in the bone marrow, double-negative (CD4-/CD8-), single-positive (CD4+ and CD8+), but not double-positive (CD4+/CD8+) thymocytes. In the periphery, mature T cells express low levels of CD127. Binding of SB/199 blocks the binding of A7R34, another antibody which recognizes mouse CD127.Applications Reported: This eBioSB/199 (SB/199) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioSB/199 (SB/199) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD127
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P16872
CD127, IL-7Ralpha, MGC107557
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBioSB/199 (SB/199)
PE
Il7r
Conjugated
Il7r
IgG2b, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16197
Videos

For Research Use Only.