missing translation for 'onlineSavingsMsg'
Learn More

CD127, APC-eFluor 780, clone: A7R34, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 47-1271-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15391640

  • £95.25 / 25µg


EU Stock 1
Add to basket

Description

Description

Description: The A7R34 monoclonal antibody reacts with mouse CD127, the high affinity alpha subunit of the mouse IL-7 receptor. IL-7 receptor alpha chain is expressed by immature B cells in the bone marrow, double-negative (CD4-CD8-), single-positive (CD4+ and CD8+), but not double-positive (CD4+CD8+) thymocytes. In the periphery, mature T cells express CD127 at low level. A7R34 inhibits binding of IL-7 to its receptor and has been used in in vivo and in vitro studies to elucidate the role of IL-7 in T and B cell development and activation. Binding of A7R34 blocks the binding of SB/199, another antibody which recognizes mouse CD127.Applications Reported: This A7R34 antibody has been reported for use in flow cytometric analysis.Applications Tested: This A7R34 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.APC-eFluor® emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD127
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
P16872
CD127, IL-7Ralpha, MGC107557
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
A7R34
APC-eFluor 780
Il7r
Conjugated
Il7r
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
16197
Videos

For Research Use Only.