missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD127 / IL-7Ra Rat, V450, Clone: A7R34, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB55-V450-100ug
Additional Details : Weight : 0.01000kg
Specifications
CD127 / IL-7Ra | |
A7R34 | |
V450 | |
NP_032385 | |
Rat | |
Protein A/G Purified | |
RUO | |
Primary | |
16172 |
Flow Cytometry | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Il17ra | |
IgG2a kappa | |
100 μg | |
Store at 2-4°C. | |
Monoclonal |