missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD127 / IL-7Ra Rat, FITC, Clone: A7R34, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB55-F-100ug
Additional Details : Weight : 0.01000kg
Specifications
CD127 / IL-7Ra | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Il17ra | |
Protein A/G Purified | |
RUO | |
16172 | |
IgG2a κ |
Flow Cytometry | |
A7R34 | |
FITC | |
NP_032385 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |