missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD123 Mouse anti-Human, PerCP-Cyanine5.5, Clone: 6H6, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 45-1239-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
IL-3Ra (Interleukin-3 receptor subunit alpha) belongs to the type I cytokine receptor family and is a heterodimer with a unique alpha chain paired with the common beta (beta c or CDw131) subunit. IL-3Ra is responsible for the transmission of signal of Interleukin-3. IL-3Ra and IL-3Rb chain heterodimerization is induced by Interleukin-3, which is required for receptor activation but not high affinity binding. IL-3Ra is also reported that IL-3Ra subunits are overexpressed on acute myeloid leukemia (AML) blasts compared with normal hematopoietic cells and are thus potential targets for novel therapeutic agents.Specifications
CD123 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P26951 | |
CD123, CDw123, SUT-1 | |
Mouse | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry | |
6H6 | |
PerCP-Cyanine5.5 | |
IL3RA | |
Liquid | |
IL3RA | |
IgG1, kappa | |
100 Tests | |
4° C, store in dark, DO NOT FREEZE! | |
Primary | |
3563 |
For Research Use Only.