missing translation for 'onlineSavingsMsg'
Learn More

CD123 Mouse anti-Human, PE-Cyanine7, Clone: 6H6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-1239-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15568466

  • £98.50 / 25 tests

Estimated Shipment
04-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. IL-3Ra (Interleukin-3 receptor subunit alpha) belongs to the type I cytokine receptor family and is a heterodimer with a unique alpha chain paired with the common beta (beta c or CDw131) subunit. IL-3Ra is responsible for the transmission of signal of Interleukin-3. IL-3Ra and IL-3Rb chain heterodimerization is induced by Interleukin-3, which is required for receptor activation but not high affinity binding. IL-3Ra is also reported that IL-3Ra subunits are overexpressed on acute myeloid leukemia (AML) blasts compared with normal hematopoietic cells and are thus potential targets for novel therapeutic agents.
Specifications

Specifications

CD123
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P26951
CD123, CDw123, SUT-1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
6H6
PE-Cyanine7
IL3RA
Liquid
IL3RA
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3563
Videos
SDS
Documents

For Research Use Only.