missing translation for 'onlineSavingsMsg'
Learn More

CD123, eFluor 450, clone: 6H6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-1239-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15519716

  • £69.57 / 25 tests


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

Description: The 6H6 monoclonal antibody reacts with human CD123, the alpha chain of the IL-3 receptor. This 60-70kDa transmembrane protein binds to IL-3 with low affinity by itself, and when associated with CD131 (common beta chain) binds IL-3 with high affinity. CD123 is expressed by myeloid precursors, macrophages, dendritic cells, mast cells, basophils, and megakaryocytes.Applications Reported: This 6H6 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 6H6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD123
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P26951
CD123, CDw123, SUT-1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
6H6
eFluor 450
IL3RA
Conjugated
IL3RA
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3563
Videos

For Research Use Only.