missing translation for 'onlineSavingsMsg'
Learn More

CD123, Biotin, clone: 6H6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-1239-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15259709

  • £43.65 / 25µg


Estimated Shipment date
from Supplier 03-04-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 6H6 monoclonal antibody reacts with human CD123, the alpha chain of the IL-3 receptor. This 60-70kDa transmembrane protein binds to IL-3 with low affinity by itself, and when associated with CD131 (common beta chain) binds IL-3 with high affinity. CD123 is expressed by myeloid precursors, macrophages, dendritic cells, mast cells, basophils, and megakaryocytes.Applications Reported: The 6H6 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 6H6 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD123
6H6
Biotin
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
'Interleukin-3 Receptor alpha, IL-3Ra
IgG1
RUO
Primary
Human
Videos