missing translation for 'onlineSavingsMsg'
Learn More

CD122, PE, clone: 5H4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1221-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15268649

  • £100.15 / 50µg


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

Description: The 5H4 antibody reacts with mouse CD122, the 90-110kDa interleukin-2 receptor beta chain. CD122 is expressed by NK cells and some T cells in the periphery and is upregulated by activation. CD122 associates with CD25 and CD132 (common gamma chain) to form the high affinity IL-2 receptor. Binding of IL-2 to CD122 is not inhibited by the 5H4 antibody.Applications Reported: The 5H4 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 5H4 antibody has been tested by flow cytometric analysis of resting and activated mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD122
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P16297
CD122, IL15RB, P70-75, CD122 antigen, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, high affinity IL-2 receptor beta subunit, high affinity IL-2 receptor subunit beta, interleukin 15 receptor, beta, interleukin-2 receptor subunit beta, p75
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
5H4
PE
Il2rb
Conjugated
Il2rb
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16185
Videos

For Research Use Only.