missing translation for 'onlineSavingsMsg'
Learn More

CD122, FITC, clone: TM-b1 (TM-beta1), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1222-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15298169

  • £56.04 / 50µg


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The TM-b1 monoclonal antibody reacts with mouse CD122, the 90-110kDa interleukin-2 receptor beta chain (IL-2R beta). CD122 is expressed by NK cells and some T cells in the periphery, and is upregulated by activation. CD122 associates with CD25 and CD132 (common gamma chain) to form the high affinity IL-2 receptor. High affinity binding of IL-2 to the IL-2 receptor is inhibited by the TM-b1 antibody.Applications Reported: This TM-b1 (TM-beta1) antibody has been reported for use in flow cytometric analysis.Applications Tested: This TM-b1 (TM-beta1) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD122
TM-b1 (TM-beta1)
FITC
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-2 Receptor beta, IL-2Rb
IgG2b
RUO
Primary
Murine
Videos