missing translation for 'onlineSavingsMsg'
Learn More

CD122, FITC, clone: TU27, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1228-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516486

  • £74.97 / 25 tests


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This TU27 monoclonal antibody reacts with human CD122, which is also known as interleukin-2 receptor beta (IL-2R beta). This receptor is expressed on NK cells, activated T cells, some B cells, monocytes, and T cell leukemias. CD122 can also be detected during early NK cell commitment. IL-2R beta associates with the IL-2R alpha (CD25) or IL-15R alpha and common gamma chain (CD132) to form the high affinity IL-2 and IL-15 receptors, respectively. By itself, this membrane receptor can bind IL-2, but with intermediate affinity. The binding of IL-2 results in internalization of the cytokine, tyrosine phosphorylation of IL-2Rbeta, and activation of signaling cascades such as the JAK/STAT pathway.Applications Reported: This TU27 antibody has been reported for use in flow cytometric analysis.Applications Tested: This TU27 antibody has been pre-titrated and tested by flow cytometric analyis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD122
TU27
FITC
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-2 Receptor beta, IL-2Rb, IL-2Rp75
IgG1
RUO
Primary
Human
Videos