missing translation for 'onlineSavingsMsg'
Learn More

CD120b (TNF Receptor II) Monoclonal Antibody (TR75-54), eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-1203-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15267187

  • £222.00 / 500µg
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The TR75-54 monoclonal antibody reacts with mouse Tumor Necrosis Factor Receptor II (TNFRII, TNFR-p80, TNFRSF1B, CD120b). TNFRII is expressed in a variety of cell types and strongly expressed on stimulated T and B lymphocytes. TNFRII is the main TNF receptor found on circulating T cells and is the major mediator of autoregulatory apoptosis in CD8+ cells. The soluble TNF receptors are truncated forms of cell surface receptors with neutralizing activity on both TNF-alpha and TNF-beta. It has been suggested that the proinflammatory and immunosuppressive properties of TNF segregate at the level of its receptors and that the pool of TNF bound to soluble receptors could represent a reservoir to control the TNF release. Applications Reported: This TR75-54 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and ELISA. Applications Tested: The TR75-54 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse TNFRII in combination with the biotin TR75-32 (cat. 13-1204) antibody for detection and recombit mouse TNFRII as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL. A standard curve consisting of doubling dilutions of the recombit standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE.

TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.
Specifications

Specifications

CD120b (TNF Receptor II)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P25119
Tnfrsf1b
Affinity chromatography
RUO
21938
4° C
Liquid
ELISA, Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot
TR75-54
Unconjugated
Tnfrsf1b
CD120b; Etanercept; p75; p75 TNF receptor; p75 TNFR; p75TNFR; p80 TNF-alpha receptor; sCD120b; soluble CD120b; soluble TNFR1B variant 1; soluble TNFR2; sTNF R2; sTNFR 2; sTNFR2; TBP-2; TBPII; TNF receptor beta chain; TNF receptor superfamily member 1B; TNF-alphaR2; TNFalpha-R2; TNFBR; TNFR; TNFR II; Tnfr-1; TNFR1B; TNFR2; TNF-R2; Tnfr-2; TNF-R75; TNFR80; TNFRII; TNF-RII; TNF-R-II; TNFR-II; TNFRSF1B; tumor necrosis factor beta receptor; tumor necrosis factor binding protein 2; Tumor necrosis factor receptor; tumor necrosis factor receptor 2; Tumor necrosis factor receptor superfamily member 1B; Tumor necrosis factor receptor superfamily member 1b, membrane form; tumor necrosis factor receptor superfamily, member 1B; tumor necrosis factor receptor type II; Tumor necrosis factor-binding protein 2
Armenian Hamster
500 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.