missing translation for 'onlineSavingsMsg'
Learn More

CD11c Mouse anti-Human, PE-Cyanine5.5, Clone: 3.9, eBioscience™

Mouse Monoclonal Antibody

£80.50 - £193.00

Specifications

Antigen CD11c
Clone 3.9
Target Species Human
Conjugate PE-Cyanine5.5
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15598846 View Documents Affymetrix eBioscience
35-0116-41
25 Tests £80.50
25 tests View alternative products
Save Up to 
»
Estimated Shipment
14-07-2020
Please to purchase this item. Need a web account? Register with us today!
15508856 View Documents Affymetrix eBioscience
35-0116-42
100 Tests £193.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment
14-07-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 690 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.
Specifications

Specifications

CD11c
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
3.9
PE-Cyanine5.5
Flow Cytometry
Primary
Monoclonal
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.