missing translation for 'onlineSavingsMsg'
Learn More

CD11c Mouse anti-Human, PE-Cyanine5.5, Clone: 3.9, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 35-0116-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15598846

  • £80.50 / 25 tests

Estimated Shipment
04-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 695 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.
Specifications

Specifications

CD11c
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P20702
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
3.9
PE-Cyanine5.5
ITGAX
Liquid
ITGAX
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3687
Videos
SDS
Documents

For Research Use Only.