missing translation for 'onlineSavingsMsg'
Learn More

CD11c, PE, clone: 3.9, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0116-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15506716

  • £146.84 / 100 tests


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 3.9 monoclonal antibody reacts with human CD11c, the 150kDa integrin alphaX chain. CD11c non-covalently associates with beta2 integrin to form the CD11c/CD18 heterodimer. This complex is expressed on monocytes, granulocytes, macrophages, NK, dendritic cells and subset of T and B lymphocytes. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions.Applications Reported: The 3.9 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 3.9 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD11c
3.9
PE
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (1μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
'Integrin alpha X, Integrin aX, ITGAX
IgG1
RUO
Primary
Human
Videos