Learn More
CD11c, FITC, clone: BU15, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD11C01
Code : 50
Additional Details : Weight : 0.50000kg
Description
ITGAX is the integrin alpha X chain protein. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles.
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.Specifications
CD11c | |
Monoclonal | |
FITC | |
ITGAX | |
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X | |
Mouse | |
Purified | |
ASR, ASR/IVD | |
3687 | |
Store at 2 - 8 °C. | |
Liquid |
Flow Cytometry | |
BU15 | |
PBS with BSA and 0.1% sodium azide | |
P20702 | |
ITGAX | |
Dendritic cells of synovial fluid | |
0.5 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |