missing translation for 'onlineSavingsMsg'
Learn More

CD11c, Biotin, clone: 3.9, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0116-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15219459

  • £49.99 / 25µg


Estimated Shipment date
from Supplier 27-07-2018
Add to basket

Description

Description

Description: The 3.9 monoclonal antibody reacts with human CD11c, the 150kDa integrin alphaX chain. CD11c non-covalently associates with beta2 integrin to form the CD11c/CD18 heterodimer. This complex is expressed on monocytes, granulocytes, macrophages, NK, dendritic cells and subset of T and B lymphocytes. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions.Applications Reported: The 3.9 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 3.9 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD11c
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P20702
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
3.9
Biotin
ITGAX
Conjugated
ITGAX
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
3687
Videos

For Research Use Only.