Learn More
CD11c Monoclonal Antibody (N418), eFluor™ 660, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
£82.00 - £201.00
Specifications
Antigen | CD11c |
---|---|
Clone | N418 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15351850
|
Affymetrix eBioscience
50-0114-80 |
25 μg |
£82.00
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15361850
|
Affymetrix eBioscience
50-0114-82 |
100 μg |
£201.00
100µg |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The N418 monoclonal antibody reacts with mouse CD11c, the integrin alpha X. CD11c non-covalently associates with beta 2 integrin to form the CD11c/CD18 heterodimer. CD11c is expressed by dendritic cells, a subset of Intestinal Intraepithelial Lymphocytes (IEL) and some activated T cells. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions. N418 binds to CD11c on splenic dendritic cells in the T-dependent areas of mouse spleen and precipitates a 150, 90 kDa heterodimer. Applications Reported: This N418 antibody has been reported for use in flow cytometric analysis. Applications Tested: This N418 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow Cytometry.Specifications
CD11c | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6 | |
Itgax | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Itgax |
N418 | |
Flow Cytometry | |
eFluor 660 | |
Armenian Hamster | |
Mouse | |
Q9QXH4 | |
16411 | |
IgG | |
Affinity chromatography | |
Antibody |
For Research Use Only.