missing translation for 'onlineSavingsMsg'
Learn More

CD11c, clone: N418, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0114-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15246807

  • £68.57 / 100µg


EU Stock 1
Add to basket

Description

Description

Description: The N418 monoclonal antibody reacts with mouse CD11c, the integrin alphaX. CD11c non-covalently associates with beta2 integrin to form the CD11c/CD18 heterodimer. CD11c is expressed by dendritic cells, a subset of Intestinal Intraepithelial Lymphocytes (IEL) and some activated T cells. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions. N418 binds to CD11c on splenic dendritic cells in the T-dependent areas of mouse spleen and precipitates a 150, 90kDa heterodimer.Applications Reported: The N418 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining of frozen tissue sections.Applications Tested: The N418 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD11c
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9QXH4
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Immunoprecipitation
N418
Unconjugated
Itgax
Purified
Itgax
IgG
100μg
Store at 2-8°C.
Primary
16411
Videos

For Research Use Only.