missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11b Monoclonal Antibody (M1/70), APC-Cyanine7, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A15390
Code : NEW
Additional Details : Weight : 0.00280kg
Description
CD11b Monoclonal antibody specifically detects CD11b in Baboon, Human, Mouse, Rabbit, Rhesus Monkey samples. It is validated for Flow Cytometry.Specifications
CD11b | |
Monoclonal | |
0.1 mg/mL | |
PBS with proprietary stabilizer and <0.1% sodium azide | |
P05555, P11215 | |
ITGAM, LOC100351865 | |
C57BL/10 splenic T cells and concanavalin A stimulated C57BL/10 splenocytes. | |
100 μg | |
Primary | |
Baboon, Human, Mouse, Rabbit, Rhesus Monkey | |
Antibody | |
IgG2b κ |
Flow Cytometry | |
M1/70 | |
APC-Cyanine7 | |
ITGAM | |
antigen CD11b (p170); antigen CD11b (p170); macrophage antigen alpha polypeptide; CD11 antigen-like family member B; CD11b; CD11B (p170); CD11b/CD18; cell surface glycoprotein MAC-1 alpha subunit; cell surface glycoprotein MAC-1 subunit alpha; complement component 3 receptor 3 subunit; complement component receptor 3 alpha-a; complement receptor type 3; CR3; CR-3 alpha chain; CR3A; F730045J24Rik; integrin alpha M; integrin alpha-M; integrin subunit alpha M; integrin, alpha M; integrin, alpha M (complement component 3 receptor 3 subunit); ITGAM; Leukocyte adhesion receptor MO1; leukocyte integrin alpha-M chain; LOC100351865; Ly-40; MAC1; Mac-1; Mac-1 alpha; Mac-1 alpha (Mac1A); MAC-1 alpha subunit; MAC1A; Mac-1a; macrophage antigen alpha; macrophage antigen alpha polypeptide; MGC117044; MO1A; Neutrophil adherence receptor; neutrophil adherence receptor alpha-M subunit; SLEB6; unnamed protein product | |
Rat | |
purified | |
RUO | |
100351865, 16409, 3684, 714832 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |