missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11b Mouse, APC, Clone: ICRF44, enQuireBio™
Mouse Monoclonal Antibody
£89.95 - £237.00
Specifications
Antigen | CD11b |
---|---|
Clone | ICRF44 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951728
|
EnquireBio
QAB25-APC-25TESTS |
25 Tests |
£89.95
25 tests |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941728
|
EnquireBio
QAB25-APC-100TESTS |
100 Tests |
£237.00
100 tests |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD11b | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
3684 | |
IgG1 κ | |
Protein A/G Purified |
ICRF44 | |
Flow Cytometry | |
APC | |
RUO | |
XP_006721108 | |
ITGAM | |
Primary | |
Store at 2-4°C. |