missing translation for 'onlineSavingsMsg'
Learn More

CD11b (activation epitope) Mouse anti-Human, PE, Clone: CBRM1/5, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0113-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15586706

  • £70.50 / 25 tests

Estimated Shipment
30-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD11b (integrin alpha-M, ITGAM, integrin alpha-X, ITGAX) is a 165 kDa adhesion molecule that associates non-covalently with integrin beta-2 (CD18). The CD11b/CD18 heterodimeric complex is also known as integrin alpha-M beta-2, Mac-1, and CR3 (complement receptor 3). CD11b is expressed on the surface of monocytes/macrophages, granulocytes, activated lymphocytes, a subset of NK cells, a subset of dendritic cells, and microglia in the brain. CD11b/CD18 functions as the receptor for ICAM-1 (CD54), ICAM-2 (CD102), ICAM-4 (CD242), CD14, CD50, CD23, heparin, iC3b, fibrinogen, and Factor X -these adhesions are critical for cell-cell and cell-matrix interactions. CD11b is expressed on 8% of spleen cells, 44% of bone marrow cells, and less than 1% of thymocytes, and is commonly used as a microglial marker in nervous tissue. The expression of CD11b increases during monocyte maturation and expression levels vary on tissue macrophages. Further, peritoneal macrophages are reported to express higher levels of CD11b than splenic macrophages. Diseases associated with CD11b dysfunction include systemic lupus erythematosus 6 and ITGAM-related susceptibility to systemic lupus erythematosus.
Specifications

Specifications

CD11b (activation epitope)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P11215
CD11B; CR3A; MAC-1; MAC1A; MGC117044; MO1A; SLEB6; Integrin alpha M; HCD11B
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
CBRM1/5
PE
ITGAM
Liquid
ITGAM
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3684
Videos
Documents

For Research Use Only.