missing translation for 'onlineSavingsMsg'
Learn More

CD11b, Biotin, clone: ICRF44, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0118-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15239459

  • £49.60 / 25µg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: The ICRF44 monoclonal antibody reacts with human CD11b, a 165kDa adhesion molecule. CD11b associated with integrin beta2 (CD18) is expressed on the surface of monocytes, granulocytes, activated lymphocytes and a subset of NK cells. CD11b is a receptor for intercellular adhesion molecule family members CD54, CD102 and CD50 as well as for iC3b. These adhesions are crucial in cell-cell and cell-matrix interactions.Applications Reported: The ICRF44 antibody has been reported for use in flow cytometric analysis.Applications Tested: The ICRF44 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD11b
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P11215
CD11B, CR3A, MAC-1, MAC1A, MGC117044, MO1A, SLEB6, Integrin alpha M
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Rhesus
Flow Cytometry
ICRF44
Biotin
ITGAM
Conjugated
ITGAM
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
3684
Videos

For Research Use Only.