missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11b (activation epitope), PerCP-eFluor™ 710, clone: CBRM1/5, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-0113-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
The CBRM1/5 monoclonal antibody reacts with an activation-specific epitope of human Mac-1. CBRM1/5 binds a subset of Mac-1 molecules on neutrophils and monocytes after stimulation with chemoattractants or phorbol esters but does not recognize Mac-1 on resting myeloid cells. Through interactions with its ligands, Mac-1 participates in adhesive cell interactions. The epitope recognized by this mAb localizes to the I domain on the \alpha chain of Mac-1 very close to the ligand binding site in a region that is widely exposed. CBRM1/5 blocks Mac-1 dependent adhesion to fibrinogen and ICAM-1 and inhibits chemoattractant-stimulated adhesion of eosinophils to the Intercellular Adhesion Molecule-1 (ICAM-1). It should be noted that low level activation may occur during processing of freshly drawn blood. Therefore the CBRM1.5 antibody may exhibit some binding to Mac-1 in these unstimulated samples. However, higher levels of Mac-1 expression are observed in activated samples when compared to unstimulated cells.Specifications
CD11b (activation epitope) | |
CBRM1/5 | |
PerCP-eFluor 710 | |
Conjugated | |
Murine | |
25 Tests | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Monoclonal |
Flow Cytometry | |
5μL (0.125μg)/test | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
Integrin alpha M, Mac-1 alpha, Complement Receptor 3 alpha (CR3A) | |
IgG1, kappa | |
RUO | |
Primary | |
Human |
For Research Use Only.