missing translation for 'onlineSavingsMsg'
Learn More

CD11a (LFA-1alpha) Rat anti-Mouse, PE-Cyanine7, Clone: M17/4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-0111-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15588346

  • £65.00 / 25µg

Estimated Shipment
30-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD11a (LFA-1 alpha, ITGAL) together with CD18 constitute leukocyte function-associated antigen 1 (LFA-1), the alphaLbeta2 integrin. CD11a is implicated in activation of LFA-1 complex. CD11a plays a central role in leukocyte intercellular adhesion through interactions with its ligands, ICAMs 1-3 (intercellular adhesion molecules 1 through 3), and also functions in lymphocyte costimulatory signaling. CD11a is expressed on the plasma membrane of leukocytes in a low-affinity conformation. Cell stimulation by chemokines or other signals leads to induction the high-affinity conformation, which supports tight binding of CD11a to its ligands, the intercellular adhesion molecules ICAM-1, -2, -3. CD11a is thus involved in interaction of various immune cells and in their tissue-specific settlement, but participates also in control of cell differentiation and proliferation and of T-cell effector functions. Blocking of CD11a function by specific antibodies or small molecules has become an important therapeutic approach in treatment of multiple inflammatory diseases. For example, humanized anti- CD11a antibody Efalizumab (Raptiva) is being used to interfere with T cell migration to sites of inflammation, binding of cholesterol-lowering drug simvastatin to CD11a allosteric site leads to immunomodulation and increase in lymphocytic cholinergic activity. Two transcript variants encoding different isoforms of CD11a have been found. Diseases associated with CD11a dysfunction include benign cephalic histiocytosis and leukocyte adhesion deficiency.
Specifications

Specifications

CD11a (LFA-1alpha)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P24063
CD11 antigen-like family member A, LFA-1 alpha, LFA-1A, antigen CD11A p180, lymphocyte function-associated antigen 1 alpha polypeptide, integrin alpha-L
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
M17/4
PE-Cyanine7
Itgal
Liquid
Itgal
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16408
Videos
Documents

For Research Use Only.