missing translation for 'onlineSavingsMsg'
Learn More

CD11a (LFA-1alpha), FITC, clone: M17/4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0111-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15258019

  • £49.20 / 50µg


EU Stock 4
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The M17/4 monoclonal antibody reacts with mouse CD11a, the 180kDa integrin alpha L, also known as the lymphocyte function associated antigen-1 (LFA-1) alpha chain. LFA-1, formed by non-covalent association of CD11a with CD18 (integrin beta 2), serves as an important adhesion molecule involved in lymphocyte and granulocyte function. CD54 (ICAM-1), CD102 (ICAM-2), and CD50 (ICAM-3) are ligands for LFA-1. CD11a is expressed by all leukocytes.Applications Reported: The M17/4 antibody has been reported for use in flow cytometric analysis.Applications Tested: The M17/4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD11a
M17/4
FITC
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Integrin alpha L, Lymphocyte Function-Associated Antigen
IgG2a
RUO
Primary
Murine
Videos