missing translation for 'onlineSavingsMsg'
Learn More

CD119 (IFN gamma Receptor 1) Monoclonal Antibody (2E2), Biotin, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-1191-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15507326

  • £106.00 / 100µg
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The monoclonal antibody 2E2 recognizes mouse CD119 also known as IFN gamma receptor1. CD119 heterodimerizes with IFN gamma receptor 2 (AF-1) forming the Class II cytokine receptor. The ligand IFN gamma binds IFN gamma receptor 1 subunit but signal transduction transpires via the IFN gamma receptor 2 subunit. The receptor is involved in host defense and expression is broad including B cells, NK cells, macrophages and dendritic cells as well as other non-hematopoietic cell types. Applications Reported: This 2E2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2E2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.
Specifications

Specifications

CD119 (IFN gamma Receptor 1)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P15261
IFNGR1
Affinity chromatography
RUO
15979
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
2E2
Biotin
IFNGR1
antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; Ifgr; IFN-gamma receptor 1; IFN-gammaR; IFN-gamma-R1; IFN-gamma-R-alpha; Ifngr; IFNGR1; IMD27A; IMD27B; immune interferon receptor 1; INF-g receptor; interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1; interferon-gamma receptor alpha chain; Nktar
Armenian Hamster
100 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.