missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117 Mouse anti-Human, BB700, Clone: YB5.B8, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566548
Additional Details : Weight : 0.01000kg
Description
CD117 Monoclonal antibody specifically detects CD117 in Human samples. It is validated for Flow Cytometry.Specifications
CD117 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
YB5.B8 | |
Mouse | |
RUO | |
Human | |
Purified |