Learn More
CD117 (c-Kit) Monoclonal Antibody (104D2), Super Bright™ 645, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£145.00 - £294.00
Specifications
Antigen | CD117 (c-Kit) |
---|---|
Clone | 104D2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943634
|
Affymetrix eBioscience
64-1178-41 |
25 Tests |
£145.00
Pack of 25 |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953634
|
Affymetrix eBioscience
64-1178-42 |
100 Tests |
£294.00
Pack of 100 |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 6H6 monoclonal antibody reacts with human CD123, the alpha chain of the IL-3 receptor. This 60-70 kDa transmembrane protein binds to IL-3 with low affinity by itself, and when associated with CD131 (common beta chain) binds IL-3 with high affinity. CD123 is expressed by myeloid precursors, macrophages, dendritic cells, mast cells, basophils, and megakaryocytes. Applications Reported: This 6H6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 6H6 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg/mL) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation.
CD117 (c-Kit) Monoclonal antibody specifically detects CD117 (c-Kit) in Human samples. It is validated for Flow Cytometry.Specifications
CD117 (c-Kit) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P10721 | |
KIT | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
KIT |
104D2 | |
Flow Cytometry | |
Super Bright 645 | |
Mouse | |
Human | |
3815 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |