missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ CD117/c-Kit/SCF-Receptor Ab-6 Rabbit Polyclonal Antibody with BSA and azide
Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD117/c-Kit/SCF-Receptor Ab-6, Rabbit Polyclonal Antibody.
£131.58
Specifications
Antigen | CD117/c-Kit/SCF-Receptor Ab-6 |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
12662987
|
Epredia™
RB-1518-P0 |
Purified | 4mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
c-kit is a transmembrane receptor tyrosine kinase. It binds to stem cell factor (SCF). It is expressed in many tissues and cells. C-kit/SCf is involved in development of several lineages of stem cells, such as germ cells, neural crest-derived melanocytes and hematopoietic precursor cells Antibody to CD117/c-kit may be helpful in the histopathological identification of gastrointestinal stromal tumors (GIST).Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: C-terminal
Immunogen: A synthetic peptide from C-terminus of human CD117/c-kit protein
Molecular Weight: 145kDa
Positive Control: Tonsil, GI Stromal Tumor (GIST)
Cellular Localization: Membrane/cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Note:
USA: RUO; Int'l: RUO
Specifications
CD117/c-Kit/SCF-Receptor Ab-6 | |
Polyclonal | |
Rabbit | |
Human | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
A synthetic peptide from C-terminus of human CD117/c-kit protein | |
Primary |