missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117 / c-Kit Rat, PE-Cy7, Clone: ACK2, enQuireBio™
Rat Monoclonal Antibody
£71.60 - £182.00
Specifications
Antigen | CD117 / c-Kit |
---|---|
Clone | ACK2 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962188
|
EnquireBio
QAB54-PE7-25UG |
25 μg |
£71.60
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952188
|
EnquireBio
QAB54-PE7-100UG |
100 μg |
£182.00
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD117 / c-Kit | |
0.2mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
16590 | |
IgG2b κ | |
Protein A/G Purified |
ACK2 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
NP_066922 | |
Kit | |
Primary | |
Store at 2-4°C. |