missing translation for 'onlineSavingsMsg'
Learn More

CD117 (c-Kit), PE, clone: 2B8, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1171-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15298639

  • £56.56 / 50µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 2B8 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor, and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145-kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1 low) and negative for lineage markers. The interaction of the mouse c-Kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells.Applications Reported: The 2B8 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 2B8 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD117 (c-kit)
2B8
PE
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Steel Factor Receptor
IgG2b
RUO
Primary
Porcine
Videos