missing translation for 'onlineSavingsMsg'
Learn More

CD117 (c-kit) Mouse anti-Human, Biotin, Clone: YB5.B8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-1179-80

UNSPSC : 12352200

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15249699

  • £48.31 / 25µg


Estimated Shipment date
from Supplier 04-07-2017
Add to basket

Disclaimers:

For Research Use Only.


Specifications

Description

The YB5.B8 monoclonal antibody reacts with human CD117, also known as c-Kit, Steel factor receptor and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145kDa molecule is expressed by hematopoietic progenitor cell subsets and mast cells. The interaction of c-Kit and Steel factor promotes proliferation and differentiation of hematopoietic progenitor cells and mast cell differentiation. CD117 is also expressed by melanocytes and plays a role in signaling and activation of these cells.

When interested in staining HSCs from peripheral blood, we recommend the use of clone 104D2. Please refer to cat. 17-1178.
Description & Specifications

Specifications

CD117 (c-kit)
YB5.B8
Biotin
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
Conjugated
cKit, Mast/Stem Cell Growth Factor Receptor
IgG1
RUO
Primary
Human