missing translation for 'onlineSavingsMsg'
Learn More

CD117 (c-Kit) Mouse anti-Human, APC, Clone: YB5.B8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1179-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15597986

  • £91.50 / 25 tests

Estimated Shipment
04-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

c-Kit, also known as CD117 and stem cell factor receptor, is a 145 kDa transmembrane tyrosine kinase encoded by the c-Kit proto-oncogene. c-Kit acts to regulate a variety of biological responses including cell proliferation, apoptosis, chemotaxis and adhesion. Ligand binding to the extracellular domain leads to autophosphorylation on several tyrosine residues within the cytoplasmic domain, and activation. c-Kit mutations correlate with tumor growth and progression in a variety of cancers including mast cell disease, gastrointestinal stromal tumor, acute myeloid leukemia, Ewing sarcoma, and lung cancer. Phosphorylation at tyrosine 703 of c-Kit allows binding of Grb2 and activation of the Ras-Raf-ERK1&2 signaling pathway.
Specifications

Specifications

CD117 (c-kit)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P10721
C-Kit, CD117, PBT, SCFR, mast/stem cell growth factor receptor Kit, p145 c-kit, piebald trait protein, proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, soluble KIT variant 1, tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein, ckit
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
YB5.B8
APC
KIT
Liquid
KIT
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3815
Videos
SDS
Documents

For Research Use Only.