missing translation for 'onlineSavingsMsg'
Learn More

CD116 Mouse anti-Human, PE, Clone: 4H1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1169-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15506866

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The 4H1 monoclonal antibody reacts with the human CD116 molecule, the α subunit of GM-CSF receptor. The α subunit associates with the common β chain (CD131) to form the high affinity receptor for GM-CSF. The GM-CSFR α chain is expressed by granulocytes, monocytes, endothelial cells, fibroblasts and some tumor cells.
Specifications

Specifications

CD116
4H1
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
GM-CSF Receptor alpha, CSF2RA
IgG1, kappa
RUO
Primary
Human
Videos

For Research Use Only.