missing translation for 'onlineSavingsMsg'
Learn More

CD116, FITC, clone: 4H1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1169-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15576476

  • £53.00 / 25 tests

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The 4H1 monoclonal antibody reacts with the human CD116 molecule, the alpha subunit of GM-CSF receptor. The alpha subunit associates with the common beta chain (CD131) to form the high affinity receptor for GM-CSF. The GM-CSFR alpha chain is expressed by granulocytes, monocytes, endothelial cells, fibroblasts and some tumor cells.Applications Reported: 4H1 has been reported for use in flow cytometric analysis.Applications Tested: This 4H1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD116
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P15509
CD116, CDw116, CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF-R-alpha, GMCSFR, GMR, MGC3848, MGC4838
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
4H1
FITC
CSF2RA
Liquid
CSF2RA
IgG1, kappa, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
1438
Videos
Documents

For Research Use Only.