missing translation for 'onlineSavingsMsg'
Learn More

CD115 (c-fms), clone: AFS98, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-1152-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15237177

  • £237.85 / 500µg


Estimated Shipment
02-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The AFS98 monoclonal antibody reacts with the mouse CD115 molecule, a receptor for macrophage colony stimulating factor (M-CSF) or colony stimulating factor-1 (CSF-1). CD115 is expressed by monocyte, macrophage, osteoclast, and some epithelial cells. It is a 150kDa c-fms gene product and belongs to immunoglobulin family. CSF-1 signaling through CSF-1R regulates the proliferation and differentiation of cells in the monocytic lineage.Applications Reported: The AFS98 antibody has been reported for use in flow cytometric analysis, immunoblotting (WB), and immunohistochemical staining of frozen tissue sections. It has also been reported in blocking of ligand binding. (Please use Functional Grade purified AFS98, cat. 16-1152, in functional assays.).Applications Tested: The AFS98 antibody has been tested by blocking of fluorochrome conjugated AFS98 in flow cytometric analysis of peritoneal exudate cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD115
AFS98
Unconjugated
Purified
Rat
500μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
FMS, Colony-Stimulating Factor 1 Receptor, M-CSF Receptor
IgG2a
RUO
Primary
Murine
Videos