missing translation for 'onlineSavingsMsg'
Learn More

CD112 (Nectin-2), APC, clone: R2.477, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1128-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15557986

  • £68.96 / 25 tests


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This R2.477 monoclonal antibody reacts with human CD112 (also known as Nectin-2 or Poliovirus Receptor Related 2), a 60-65kDa immunoglobulin-like transmembrane cell adhesion molecule. This cell surface molecule is expressed on hematopoietic cells, endothelial, epithelial, as well as tumor cell lines such as Jurkat, U937, and MCF7. CD112 is a ligand for DNAM-1 (CD226), a leukocyte adhesion molecule expressed on natural killer (NK) cells. Interaction between CD112 and DNAM-1 promotes NK cytotoxic activity, leading to tumor cell lysis. In addition to its role in tumor suppression, CD112 plays a cooperative role in cell-cell adhesion, particularly in the formation of adherens junctions between neighboring epithelial cells and fibroblasts. CD112 can form homophilic and heterophilic interactions to regulate contact inhibition of cell movement and proliferation. Finally, CD112 has been reported to be a receptor for herpesvirus entry into cells.This monoclonal antibody has been reported to block cell adhesion and herpesvirus entry.Applications Reported: This R2.477 antibody has been reported for use in flow cytometric analysis.Applications Tested: This R2.477 antibody has been pre-titrated and tested by flow cytometric analysis of human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD112 (Nectin-2)
R2.477
APC
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
PVRL2, Nectin-2, PRR2
IgG1
RUO
Primary
Human
Videos