missing translation for 'onlineSavingsMsg'
Learn More

CD109 Mouse anti-Human, PE, Clone: HU17, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1099-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566856

  • £72.50 / 25 tests

Estimated Shipment
29-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD109 is a member of the alpha2-macroglobulin/complement superfamily. The encoded GPI-linked glycoprotein is found on the cell surface of platelets, activated T-cells, and endothelial cells. The protein binds to and negatively regulates signaling of transforming growth factor beta (TGF-beta).
Specifications

Specifications

CD109
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q6YHK3
CPAMD7, RP11-525G3.1, 150 kDa TGF-beta-1-binding protein
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HU17
PE
CD109
Liquid
CD109
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
135228
Videos
Documents

For Research Use Only.