missing translation for 'onlineSavingsMsg'
Learn More

CD109 Mouse anti-Human, PE, Clone: HU17, eBioscience™

Mouse Monoclonal Antibody

£152.00

Specifications

Antigen CD109
Clone HU17
Target Species Human
Conjugate PE
Isotype IgG1, kappa
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15576856
View Documents
Affymetrix eBioscience
12-1099-42
100 Tests £152.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment
04-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD109 is a member of the alpha2-macroglobulin/complement superfamily. The encoded GPI-linked glycoprotein is found on the cell surface of platelets, activated T-cells, and endothelial cells. The protein binds to and negatively regulates signaling of transforming growth factor beta (TGF-beta).
Specifications

Specifications

CD109
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
HU17
PE
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.