missing translation for 'onlineSavingsMsg'
Learn More

CD107b (LAMP-2), PE, clone: M3/84, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5989-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15229239

  • POA / 50µg

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.

Disclaimers:

For Research Use Only.


Description

Description

Description: The M3/84 monoclonal antibody reacts with mouse Mac-3, the 110kDa protein expressed by mononuclear phagocytes. The Mac-3 antigen is expressed intracellularly by bone marrow monocyte lineage and is upregulated during macrophage differentiation. Peritoneal and tissue macrophages, dendritic cells, and endothelial cells express this antigen on their surface. It is reported that Mac-3 may be identical to CD107b.Applications Reported: The M3/84 antibody has been reported for use in flow cytometric analysis.Applications Tested: The M3/84 antibody has been tested by flow cytometric analysis of mouse bone marrow and thioglycolate-elicited peritoneal exudates cells and can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD107b (Mac-3)
M3/84
PE
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
LAMP-2, LAMP2
IgG1
RUO
Primary
Murine
Videos