missing translation for 'onlineSavingsMsg'
Learn More

CD107b (LAMP-2) Rat anti-Mouse, FITC, Clone: eBioABL-93 (ABL-93), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1072-82

Code : Z2

Additional Details : Weight : 0.25000kg

Product Code. 15248169

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The eBioABL-93 monoclonal antibody reacts with mouse CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated protein of approximately 120kDa that is expressed intracellularly in lysosomal membranes. It is also transiently expressed on the surface of cytolytic T cells during degranulation, but to a lesser extent than CD107a. It has been reported that CD107b may be identical to the mouse Mac-3 antigen.
Specifications

Specifications

CD107b (LAMP-2)
eBioABL-93 (ABL-93)
FITC
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
LAMP2, lysosomal-associated membrane protein 2
IgG2a, kappa
RUO
Primary
Murine
Videos

For Research Use Only.