missing translation for 'onlineSavingsMsg'
Learn More

CD107b (LAMP-2), eFluor 660, clone: eBioABL-93 (ABL-93), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-1072-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15351880

  • £62.63 / 25µg


Estimated Shipment date
from Supplier 04-06-2018
Add to basket

Description

Description

Description: The eBioABL-93 monoclonal antibody reacts with mouse CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated protein of approximately 120kDa that is expressed intracellularly in lysosomal membranes. It is also transiently expressed on the surface of cytolytic T cells during degranulation, but to a lesser extent than CD107a. It has been reported that CD107b may be identical to the mouse Mac-3 antigen.Applications Reported: This ABL-93 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This eBioABL-93 (ABL-93) antibody has been tested by intracellular staining and flow cytometric analysis of thioglycolate elicited peritoneal mouse cells. This can be used at less than or equal to 1μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD107b (LAMP-2)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P17047
RP23-193O17.2, CD107b, Lamp-2, Lamp-2a, Lamp-2b, Lamp-2c, Mac3
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBioABL-93
eFluor 660
Lamp2
Conjugated
Lamp2
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16784
Videos

For Research Use Only.